Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERMARD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278531.1 | NP_001265460.1 | 4 | 388 | ||
NM_001278532.1 | NP_001265461.1 | 3 | 219 | |||
NM_001278533.1 | NP_001265462.1 | 4 | 388 | |||
NM_018341.2 | NP_060811.1 | 4 | 388 | |||
XM_011535938.2 | XP_011534240.1 | 4 | 388 | |||
XM_011535939.2 | XP_011534241.1 | 4 | 1478 | |||
XM_011535940.2 | XP_011534242.1 | 4 | 1287 | |||
XM_017011032.1 | XP_016866521.1 | 4 | 1473 | |||
XM_017011033.1 | XP_016866522.1 | 4 | 1282 | |||
GenBank mRNA | AK002014.1 | - | 4 | 385 | ||
AK294090.1 | - | 3 | 216 | |||
BC044812.1 | - | 4 | 372 | |||
CA396814.1 | - | 2 | 68 | |||
DA650477.1 | - | 4 | 388 |