Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERMARD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278531.1 | NP_001265460.1 | 9, 10 | 997 | ||
NM_001278532.1 | NP_001265461.1 | 8, 9 | 828 | |||
NM_001278533.1 | NP_001265462.1 | 9, 10 | 997 | |||
NM_018341.2 | NP_060811.1 | 9, 10 | 997 | |||
XM_011535939.2 | XP_011534241.1 | 9, 10 | 2087 | |||
XM_011535940.2 | XP_011534242.1 | 9, 10 | 1896 | |||
XM_017011027.1 | XP_016866516.1 | 9, 10 | 1837 | |||
XM_017011028.1 | XP_016866517.1 | 9, 10 | 1162 | |||
XM_017011029.1 | XP_016866518.1 | 9, 10 | 922 | |||
XM_017011030.1 | XP_016866519.1 | 8, 9 | 746 | |||
XM_017011031.1 | XP_016866520.1 | 8, 9 | 811 | |||
XM_017011032.1 | XP_016866521.1 | 9, 10 | 2082 | |||
XM_017011033.1 | XP_016866522.1 | 9, 10 | 1891 | |||
GenBank mRNA | AK001369.1 | - | 8, 9 | 858 | ||
AK002014.1 | - | 9, 10 | 994 | |||
AK294090.1 | - | 8, 9 | 825 | |||
BC044812.1 | - | 9, 10 | 981 |