Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSENEN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281532.1 | NP_001268461.1 | 3 | 281 | ||
NM_172341.2 | NP_758844.1 | 3 | 319 | |||
GenBank mRNA | AF220053.1 | - | 3 | 297 | ||
AK312233.1 | - | 3 | 245 | |||
BC009575.1 | - | 3 | 239 | 4096271 | ||
BE563017.1 | - | 2 | 124 | |||
BQ222622.1 | - | 303 | ||||
BU934029.1 | - | 311 | ||||
CB136623.1 | - | 3 | 319 |