Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZC4H2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001178032.2 | NP_001171503.1 | 5 | 920 | ||
NM_001178033.2 | NP_001171504.1 | 4 | 595 | |||
NM_001243804.1 | NP_001230733.1 | 5 | 665 | |||
NM_018684.3 | NP_061154.1 | 5 | 758 | |||
NR_045044.1 | 5 | 1013 | ||||
GenBank mRNA | AB032992.1 | - | 3 | 2779 | ||
AF270491.1 | - | 5 | 690 | |||
AK001547.1 | - | 2 | 187 | |||
AK022807.1 | - | 5 | 889 | |||
AK022918.1 | - | 5 | 652 | |||
AK123763.1 | - | 6 | 1151 | |||
AK222708.1 | - | 5 | 675 | |||
AK293570.1 | - | 4 | 512 | |||
AK315485.1 | - | 5 | 673 | |||
BC004411.1 | - | 5 | 709 | 3639978 | ||
CX782941.1 | - | 5 | 630 |