Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NSFL1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206736.1 | NP_001193665.1 | 7 | 1444 | ||
NM_016143.4 | NP_057227.2 | 6 | 1438 | |||
NM_018839.4 | NP_061327.2 | 5 | 1345 | |||
NR_038164.1 | 5 | 1245 | ||||
XM_006723592.2 | XP_006723655.1 | 7 | 1377 | |||
XM_006723593.2 | XP_006723656.1 | 6 | 1279 | |||
XM_006723594.2 | XP_006723657.1 | 6 | 1371 | |||
XM_011529300.1 | XP_011527602.1 | 8 | 1481 | |||
XM_011529301.1 | XP_011527603.1 | 7 | 1475 | |||
XM_011529302.2 | XP_011527604.1 | 6 | 1995 | |||
XM_011529304.1 | XP_011527606.1 | 7 | 682 | |||
XM_011529305.2 | XP_011527607.1 | 8 | 1481 | |||
XM_017027956.1 | XP_016883445.1 | 5 | 1985 | |||
XM_017027957.1 | XP_016883446.1 | 8 | 1579 | |||
XM_017027958.1 | XP_016883447.1 | 4 | 508 | |||
GenBank mRNA | AF078856.1 | - | 6 | 583 | ||
AF086909.1 | - | 7 | 590 | |||
AF283774.2 | - | 6 | 589 | |||
AK001511.1 | - | 5 | 481 | |||
AK093845.1 | - | 6 | 1438 | |||
AK223334.1 | - | 6 | 589 | |||
AK223341.1 | - | 6 | 590 | |||
AK297403.1 | - | 5 | 396 | |||
AK315433.1 | - | 6 | 589 | |||
BC002801.2 | - | 6 | 646 | 3635947 |