Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C20orf24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199534.1 | NP_001186463.1 | 4 | 650 | ||
NM_018840.4 | NP_061328.1 | 4 | 622 | |||
NM_199483.2 | NP_955777.1 | 3 | 492 | |||
NR_026562.3 | 5 | 733 | ||||
GenBank mRNA | AF112213.1 | - | 4 | 586 | ||
AF274948.1 | - | 4 | 435 | |||
BC001871.1 | - | 4 | 567 | 3546888 | ||
BC004446.1 | - | 5 | 658 | 2819807 | ||
BI223439.1 | - | 4 | 627 | |||
BQ086248.1 | - | 4 | 552 | |||
BU189639.1 | - | 3 | 404 | |||
BU194101.1 | - | 419 | ||||
BU848918.1 | - | 4 | 581 | |||
CD370956.1 | - | 1 | 503 |
Gene Symbol: TGIF2-C20orf24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199535.1 | NP_001186464.1 | 5 | 598 |