Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCDHB14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018934.3 | NP_061757.1 | 1 | 2117 | ||
GenBank mRNA | AF152493.1 | - | 1 | 937 | ||
AF217744.1 | - | 1 | 937 | |||
AK074555.1 | - | 1 | 1069 | |||
AK298296.1 | - | 2 | 716 | |||
AK301346.1 | - | 1 | 1100 | |||
BC098144.1 | - | 1 | 936 | |||
BC099728.1 | - | 1 | 937 | 40019185 |