Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCDHB13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018933.3 | NP_061756.1 | 1 | 1499 | ||
GenBank mRNA | AF152492.1 | - | 1 | 1294 | ||
AF217745.1 | - | 1 | 1294 | |||
AK021915.1 | - | 1 | 1482 | |||
AK292821.1 | - | 1 | 1461 | |||
AK296151.1 | - | 1 | 1462 | |||
AY358427.1 | - | 1 | 1464 | |||
BC033068.1 | - | 1 | 1452 | 5446016 | ||
BC051348.1 | - | 1 | 1452 | 6483735 | ||
BC071934.1 | - | 1 | 1452 | 6383780 |