Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCDHB10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018930.3 | NP_061753.1 | 1 | 2522 | ||
GenBank mRNA | AF131761.1 | - | 1 | 1345 | ||
AF152489.1 | - | 1 | 2348 | |||
AF217748.1 | - | 1 | 2348 | |||
AK056391.1 | - | 1 | 2532 | |||
AK315583.1 | - | 1 | 2542 | |||
AY358720.1 | - | 1 | 2526 | |||
BC031837.1 | - | 1 | 2522 | 4931995 |