Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERF1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022978.2 | NP_075267.1 | 3 | 981 | ||
GenBank mRNA | BC103687.1 | - | 3 | 801 | 40000408 | |
BC104738.1 | - | 3 | 799 | 40000411 |
Gene Symbol: ATF7IP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286514.1 | NP_001273443.1 | 15 | 5680 | ||
NM_018179.4 | NP_060649.3 | 15 | 5683 | |||
NM_181352.1 | NP_851997.1 | 15 | 5824 | |||
XM_005253424.3 | XP_005253481.1 | 15 | 5952 | |||
XM_006719108.3 | XP_006719171.1 | 15 | 6096 | |||
XM_006719109.3 | XP_006719172.1 | 15 | 5963 | |||
XM_011520754.2 | XP_011519056.1 | 15 | 5646 | |||
XM_011520755.2 | XP_011519057.1 | 15 | 5877 | |||
XM_017019638.1 | XP_016875127.1 | 15 | 5542 | |||
XM_017019639.1 | XP_016875128.1 | 15 | 6093 |
Gene Symbol: GRM6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX647168.1 | - | 2 | 5344 |
Gene Symbol: LOC729275 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK129559.1 | - | 1 | 1343 |
Gene Symbol: MTBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK022122.1 | - | 4 | 1906 |
Gene Symbol: MTHFD2P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_077228.1 | 3 | 927 | |||
GenBank mRNA | AK092228.1 | - | 3 | 927 | ||
BJ991572.1 | - | 3 | 363 | |||
BJ991573.1 | - | 1 | 485 |
Gene Symbol: PRMT8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB209027.1 | - | 3 | 2837 |
Gene Symbol: SERF1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021967.2 | NP_068802.1 | 3 | 987 | ||
GenBank mRNA | AF073519.1 | - | 3 | 963 | ||
BC035932.1 | - | 3 | 915 | 4701505 |