Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERF1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022978.2 | NP_075267.1 | 3 | 1007 | ||
GenBank mRNA | BC104738.1 | - | 3 | 825 | 40000411 |
Gene Symbol: SERF1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021967.2 | NP_068802.1 | 3 | 1013 | ||
GenBank mRNA | AF073519.1 | - | 3 | 989 | ||
BC035932.1 | - | 3 | 941 | 4701505 |