Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLE4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019896.2 | NP_063949.2 | 1 | 216 | ||
GenBank mRNA | AI815728.1 | - | 1 | 243 | ||
BC031331.1 | - | 1 | 211 | 5443810 | ||
BU597500.1 | - | 223 | ||||
CA454961.1 | - | 1 | 158 |