Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206646.1 | NP_001193575.1 | 2 | 165 | ||
NM_001206647.1 | NP_001193576.1 | 2 | 165 | |||
NM_001206648.1 | NP_001193577.1 | 2 | 165 | |||
NM_020642.3 | NP_065693.2 | 2 | 165 | |||
NR_045417.1 | 1 | 347 | ||||
NR_045418.1 | 1 | 347 | ||||
XM_017018011.1 | XP_016873500.1 | 2 | 185 | |||
XM_017018012.1 | XP_016873501.1 | 2 | 193 | |||
GenBank mRNA | AF493783.1 | - | 2 | 165 | ||
AF493784.1 | - | 2 | 165 | |||
AF493785.1 | - | 2 | 165 | |||
AF512007.1 | - | 1 | 89 | |||
AK075308.1 | - | 284 | ||||
AK294506.1 | - | 1 | 338 | |||
AK294549.1 | - | 1 | 326 | |||
AW340643.1 | - | 1 | 20 | |||
BC030996.2 | - | 1 | 95 | 4609949 | ||
BM706322.1 | - | 1 | 201 | |||
BP355293.1 | - | 1 | 347 |