Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193557.1 | NP_001180486.1 | 3 | 213 | ||
NM_020189.5 | NP_064574.1 | 3 | 285 | |||
NR_036471.1 | 3 | 285 | ||||
NR_036472.1 | 3 | 285 | ||||
GenBank mRNA | AF173296.1 | - | 3 | 355 | ||
AF201940.1 | - | 3 | 300 | |||
AK000487.1 | - | 3 | 189 | |||
AK095651.1 | - | 3 | 284 | |||
AK290752.1 | - | 3 | 229 | |||
AK316560.1 | - | 3 | 280 | |||
BC007870.2 | - | 3 | 201 | 4303290 | ||
BX648621.1 | - | 1 | 1034 | |||
CR457183.1 | - | 2 | 139 | |||
DA548665.1 | - | 3 | 159 | |||
DA762166.1 | - | 3 | 267 | |||
DB168034.1 | - | 3 | 285 |