Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OLFML3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286352.1 | NP_001273281.1 | 4 | 1252 | ||
NM_001286353.1 | NP_001273282.1 | 3 | 1229 | |||
NM_020190.3 | NP_064575.1 | 3 | 1100 | |||
XM_017001848.1 | XP_016857337.1 | 3 | 1262 | |||
GenBank mRNA | AF201945.1 | - | 3 | 1034 | ||
AK075353.1 | - | 3 | 1002 | |||
AK075369.1 | - | 3 | 1048 | |||
AK075479.1 | - | 3 | 1013 | |||
AK075544.1 | - | 2 | 1331 | |||
AK092766.1 | - | 4 | 1202 | |||
AK222471.1 | - | 3 | 1017 | |||
AK223221.1 | - | 3 | 1030 | |||
AK297902.1 | - | 3 | 936 | |||
AY358954.1 | - | 3 | 1083 | |||
AY464015.1 | - | 3 | 1065 | |||
BC009920.2 | - | 3 | 1025 | 3010136 | ||
KC237062.1 | - | 3 | 974 |