Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL23AP32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC067268.1 | - | 1 | 1460 |
Gene Symbol: SPTBN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB209551.1 | - | 1 | 2792 |