Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSRP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321772.1 | NP_001308701.1 | 5 | 2084 | ||
NM_020317.4 | NP_064713.3 | 5 | 1080 | |||
NR_135143.1 | 4 | 2596 | ||||
NR_135144.1 | 5 | 1671 | ||||
NR_135777.1 | 4 | 2571 | ||||
NR_135778.1 | 5 | 1945 | ||||
NR_135780.1 | 5 | 2049 | ||||
NR_135781.1 | 6 | 1716 | ||||
NR_135782.1 | 6 | 1398 | ||||
NR_135783.1 | 6 | 1125 | ||||
NR_135784.1 | 4 | 2596 | ||||
NR_135785.1 | 6 | 1124 | ||||
NR_135786.1 | 4 | 2596 | ||||
NR_135787.1 | 6 | 2720 | ||||
NR_135788.1 | 4 | 2662 | ||||
NR_135789.1 | 4 | 3600 | ||||
XM_011541797.1 | XP_011540099.1 | 5 | 1045 | |||
GenBank mRNA | AF267856.1 | - | 4 | 2524 | ||
AK226012.1 | - | 5 | 1608 | |||
AK292532.1 | - | 4 | 2532 | |||
AL576262.3 | - | 1 | 389 | |||
BC001892.1 | - | 5 | 1004 | 3534629 | ||
BC041843.1 | - | 6 | 2631 | 5269181 | ||
BC065040.1 | - | 5 | 993 | 6147639 | ||
BM875537.1 | - | 2 | 115 |