Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSRP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321772.1 | NP_001308701.1 | 4 | 2012 | ||
NM_020317.4 | NP_064713.3 | 4 | 1008 | |||
NR_135143.1 | 3 | 2524 | ||||
NR_135144.1 | 4 | 1599 | ||||
NR_135777.1 | 3 | 2499 | ||||
NR_135778.1 | 4 | 1873 | ||||
NR_135780.1 | 4 | 1977 | ||||
NR_135781.1 | 5 | 1644 | ||||
NR_135782.1 | 5 | 1326 | ||||
NR_135783.1 | 5 | 1053 | ||||
NR_135784.1 | 3 | 2524 | ||||
NR_135785.1 | 5 | 1052 | ||||
NR_135786.1 | 3 | 2524 | ||||
NR_135787.1 | 5 | 2648 | ||||
NR_135788.1 | 3 | 2590 | ||||
NR_135789.1 | 3 | 3528 | ||||
XM_011541797.1 | XP_011540099.1 | 4 | 973 | |||
GenBank mRNA | AF267856.1 | - | 3 | 2452 | ||
AK226012.1 | - | 4 | 1536 | |||
AK292532.1 | - | 3 | 2460 | |||
AL576262.3 | - | 2 | 461 | |||
BC001892.1 | - | 4 | 932 | 3534629 | ||
BC041843.1 | - | 5 | 2559 | 5269181 | ||
BC065040.1 | - | 4 | 921 | 6147639 | ||
BM875537.1 | - | 1 | 43 |