Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGTRAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040194.1 | NP_001035284.1 | 3, 4 | 292 | ||
NM_001040195.1 | NP_001035285.1 | 2, 3 | 257 | |||
NM_001040196.1 | NP_001035286.1 | 4, 5 | 389 | |||
NM_001040197.1 | NP_001035287.1 | 4, 5 | 389 | |||
NM_020350.4 | NP_065083.3 | 3, 4 | 292 | |||
XM_011541799.2 | XP_011540101.2 | 4, 5 | 306 | |||
XM_011541800.2 | XP_011540102.1 | 4, 5 | 306 | |||
XM_011541802.1 | XP_011540104.1 | 5, 6 | 403 | |||
GenBank mRNA | AF165187.1 | - | 3, 4 | 168 | ||
AF312374.1 | - | 3, 4 | 223 | |||
AK290042.1 | - | 3, 4 | 235 | |||
BC011946.1 | - | 3, 4 | 198 | |||
BC017328.2 | - | 3, 4 | 230 | 4546585 | ||
BE782705.1 | - | 342 | ||||
BI117521.1 | - | 3, 4 | 264 | |||
CA866314.1 | - | 2, 3 | 263 | |||
CD674964.1 | - | 4, 5 | 269 |