Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCNP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320395.1 | NP_001307324.1 | 2 | 257 | ||
NM_001320397.1 | NP_001307326.1 | 2 | 257 | |||
NM_001320398.1 | NP_001307327.1 | 2 | 257 | |||
NM_001320399.1 | NP_001307328.1 | 3 | 383 | |||
NM_001320400.1 | NP_001307329.1 | 2 | 257 | |||
NM_001320401.1 | NP_001307330.1 | 2 | 257 | |||
NM_020357.2 | NP_065090.1 | 2 | 257 | |||
NR_135223.1 | 2 | 205 | ||||
NR_135225.1 | 2 | 205 | ||||
NR_135226.1 | 2 | 205 | ||||
NR_135227.1 | 2 | 458 | ||||
NR_135228.1 | 2 | 205 | ||||
NR_135230.1 | 2 | 205 | ||||
NR_135232.1 | 3 | 300 | ||||
XM_017006922.1 | XP_016862411.1 | 2 | 171 | |||
GenBank mRNA | AB037675.1 | - | 2 | 153 | ||
AK025551.1 | - | 2 | 97 | |||
AK098760.1 | - | 2 | 208 | |||
AK222978.1 | - | 2 | 148 | |||
AK314629.1 | - | 2 | 138 | |||
BC013916.1 | - | 2 | 165 | 3929908 | ||
BC022001.2 | - | 2 | 155 | 4823990 | ||
BC107796.1 | - | 2 | 99 | |||
BF244508.1 | - | 149 | ||||
BF541732.1 | - | 2 | 156 | |||
BI547939.1 | - | 2 | 370 | |||
BI596913.1 | - | 3 | 309 | |||
CD696085.1 | - | 2 | 172 | |||
CN482876.1 | - | 3 | 184 | |||
DA041324.1 | - | 2 | 113 | |||
DA304502.1 | - | 2 | 205 | |||
DA710370.1 | - | 2 | 147 | |||
DA817951.1 | - | 2 | 125 | |||
DA821754.1 | - | 2 | 119 | |||
DB060329.1 | - | 2 | 168 | |||
DB481581.1 | - | 2 | 93 |