Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RALGAPA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020343.3 | NP_065076.2 | 11, 12 | 1554 | ||
XM_005260768.2 | XP_005260825.1 | 11, 12 | 1569 | |||
XM_006723598.1 | XP_006723661.1 | 11, 12 | 1569 | |||
XM_006723599.1 | XP_006723662.1 | 11, 12 | 1569 | |||
XM_011529309.1 | XP_011527611.1 | 11, 12 | 1569 | |||
XM_011529310.2 | XP_011527612.1 | 9, 10 | 1190 | |||
XM_017027972.1 | XP_016883461.1 | 11, 12 | 1569 | |||
XM_017027973.1 | XP_016883462.1 | 8, 9 | 1032 | |||
XM_017027974.1 | XP_016883463.1 | 8, 9 | 1027 | |||
XM_017027975.1 | XP_016883464.1 | 11, 12 | 1473 | |||
XM_017027976.1 | XP_016883465.1 | 9, 10 | 1326 | |||
XM_017027977.1 | XP_016883466.1 | 11, 12 | 1569 | |||
GenBank mRNA | AK002211.1 | - | 3, 4 | 424 | ||
BC171842.1 | - | 9, 10 | 1326 | |||
DQ310704.1 | - | 11, 12 | 1411 |