Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RALGAPA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020343.3 | NP_065076.2 | 36 | 5443 | ||
XM_005260768.2 | XP_005260825.1 | 36 | 5458 | |||
XM_006723598.1 | XP_006723661.1 | 37 | 5599 | |||
XM_006723599.1 | XP_006723662.1 | 37 | 5599 | |||
XM_011529309.1 | XP_011527611.1 | 37 | 5599 | |||
XM_011529310.2 | XP_011527612.1 | 35 | 5220 | |||
XM_011529311.2 | XP_011527613.2 | 23 | 5559 | |||
XM_017027972.1 | XP_016883461.1 | 37 | 5599 | |||
XM_017027973.1 | XP_016883462.1 | 34 | 5062 | |||
XM_017027974.1 | XP_016883463.1 | 34 | 5057 | |||
XM_017027975.1 | XP_016883464.1 | 36 | 5391 | |||
XM_017027976.1 | XP_016883465.1 | 34 | 5244 | |||
GenBank mRNA | AK002211.1 | - | 28 | 4312 | ||
AK022881.1 | - | 5 | 532 | |||
AK127228.1 | - | 9 | 3614 | |||
BC013749.1 | - | 5 | 591 | |||
BC171842.1 | - | 34 | 5244 | |||
DQ310704.1 | - | 36 | 5300 |