Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THAP11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020457.2 | NP_065190.2 | 1 | 501 | ||
GenBank mRNA | AK289367.1 | - | 1 | 477 | ||
BC012182.1 | - | 1 | 463 | 4554554 | ||
BI261822.1 | - | 1 | 411 | |||
EF036502.1 | - | 1 | 129 | |||
L10378.1 | - | 1 | 338 | |||
L10379.1 | - | 1 | 338 |