Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYRM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020466.4 | NP_065199.1 | 2 | 143 | ||
NR_028493.1 | 2 | 285 | ||||
NR_028494.1 | 2 | 285 | ||||
NR_028495.1 | 2 | 153 | ||||
GenBank mRNA | AK024783.1 | - | 2 | 141 | ||
AK094994.1 | - | 2 | 194 | |||
AK311180.1 | - | 2 | 202 | |||
AK311275.1 | - | 2 | 138 | |||
AK311948.1 | - | 2 | 128 | |||
AL122090.1 | - | 2 | 126 | |||
AV710669.1 | - | 2 | 143 | |||
BC009782.1 | - | 2 | 137 | 4092710 | ||
BC104668.1 | - | 2 | 126 | 6717725 | ||
BP280644.1 | - | 2 | 143 |