Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GALNT16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168368.1 | NP_001161840.1 | 10, 11 | 1415 | ||
NM_020692.2 | NP_065743.2 | 10, 11 | 1415 | |||
XM_011537005.1 | XP_011535307.1 | 10, 11 | 1227 | |||
XM_011537006.2 | XP_011535308.1 | 10, 11 | 1227 | |||
XM_011537007.2 | XP_011535309.1 | 10, 11 | 1227 | |||
XM_017021498.1 | XP_016876987.1 | 11, 12 | 1402 | |||
GenBank mRNA | AB032956.1 | - | 10, 11 | 1205 | ||
AB078143.1 | - | 10, 11 | 1088 | |||
AJ505951.1 | - | 10, 11 | 1088 | |||
AK289745.1 | - | 10, 11 | 1174 | |||
AK315848.1 | - | 10, 11 | 1179 | |||
BC036812.1 | - | 10, 11 | 1166 | 5734009 | ||
BC098578.1 | - | 10, 11 | 1181 | 30924466 | ||
BC122545.1 | - | 10, 11 | 1188 | 40084363 |