Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIAA1143 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320334.1 | NP_001307263.1 | 3 | 469 | ||
NM_020696.3 | NP_065747.1 | 3 | 414 | |||
GenBank mRNA | AB032969.1 | - | 2 | 221 | ||
AK289549.1 | - | 3 | 386 | |||
BC008468.1 | - | 3 | 389 | 4277104 | ||
BC016790.1 | - | 3 | 348 | 4102473 | ||
BC070360.1 | - | 3 | 344 | |||
CD049061.1 | - | 3 | 411 | |||
DA183520.1 | - | 3 | 407 | |||
DA192476.1 | - | 3 | 389 | |||
DB238094.1 | - | 3 | 425 |