Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLEKHG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029884.1 | NP_001025055.1 | 16 | 1849 | ||
XM_005267064.3 | XP_005267121.1 | 15 | 3928 | |||
XM_005267065.3 | XP_005267122.1 | 17 | 1875 | |||
XM_005267066.2 | XP_005267123.1 | 15 | 2732 | |||
XM_006715521.1 | XP_006715584.1 | 17 | 2102 | |||
XM_006715522.3 | XP_006715585.1 | 15 | 3969 | |||
XM_011535981.2 | XP_011534283.1 | 15 | 4725 | |||
XM_011535982.2 | XP_011534284.1 | 14 | 4686 | |||
XM_011535983.2 | XP_011534285.1 | 16 | 3150 | |||
XM_017011102.1 | XP_016866591.1 | 14 | 2695 | |||
GenBank mRNA | AB033035.1 | - | 9 | 722 | ||
AJ420468.1 | - | 13 | 1087 | |||
AK056300.1 | - | 15 | 1748 | |||
AK293288.1 | - | 15 | 1780 | |||
AK294798.1 | - | 15 | 1903 | |||
AK307631.1 | - | 15 | 1802 | |||
BC089428.1 | - | 11 | 960 | |||
BC140864.1 | - | 14 | 1586 | 9021732 | ||
BC151134.1 | - | 14 | 1586 | 9057034 |