Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ODF2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007022.2 | NP_001007023.2 | 2 | 375 | ||
NM_001184765.1 | NP_001171694.1 | 2 | 382 | |||
NM_001184766.1 | NP_001171695.1 | 2 | 382 | |||
NM_020729.2 | NP_065780.2 | 2 | 382 | |||
XM_005271054.3 | XP_005271111.1 | 2 | 380 | |||
XM_011541815.2 | XP_011540117.1 | 2 | 380 | |||
XM_011541819.2 | XP_011540121.1 | 2 | 380 | |||
XM_011541821.2 | XP_011540123.1 | 2 | 380 | |||
XM_011541823.2 | XP_011540125.1 | 2 | 380 | |||
XM_011541824.2 | XP_011540126.1 | 2 | 380 | |||
XM_017001876.1 | XP_016857365.1 | 2 | 172 | |||
XM_017001877.1 | XP_016857366.1 | 2 | 172 | |||
XM_017001878.1 | XP_016857367.1 | 2 | 380 | |||
XM_017001879.1 | XP_016857368.1 | 2 | 380 | |||
XM_017001880.1 | XP_016857369.1 | 2 | 380 | |||
XM_017001881.1 | XP_016857370.1 | 2 | 385 | |||
GenBank mRNA | AB033055.1 | - | 2 | 295 | ||
AK302792.1 | - | 2 | 354 | |||
AK303209.1 | - | 2 | 379 | |||
BC026105.2 | - | 2 | 363 | |||
BC029420.1 | - | 2 | 100 | |||
BC047368.1 | - | 2 | 322 | 5276959 | ||
BC091490.1 | - | 2 | 281 | 30367572 | ||
BG777878.1 | - | 2 | 361 | |||
BI768058.1 | - | 2 | 337 |