Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAVS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206491.1 | NP_001193420.1 | 5 | 1069 | ||
NM_020746.4 | NP_065797.2 | 6 | 1244 | |||
NR_037921.1 | 5 | 1071 | ||||
GenBank mRNA | AB033097.1 | - | 5 | 1150 | ||
AB232371.1 | - | 5 | 1144 | |||
AK023799.1 | - | 6 | 1199 | |||
AK291785.1 | - | 5 | 1028 | |||
AK296897.1 | - | 5 | 1036 | |||
AL833736.1 | - | 1 | 404 | |||
BC044952.1 | - | 6 | 1178 | 5751684 | ||
BX649084.1 | - | 5 | 1030 | |||
DQ167126.1 | - | 5 | 1072 | |||
DQ174270.1 | - | 5 | 1072 | |||
DQ181928.1 | - | 5 | 1072 | |||
EF467323.1 | - | 4 | 897 | |||
EF467324.1 | - | 4 | 899 | |||
KC415005.1 | - | 5 | 1072 |