Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EBF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110514.1 | NP_001103984.1 | 10 | 1108 | ||
XM_006723600.1 | XP_006723663.1 | 10 | 903 | |||
XM_017027983.1 | XP_016883472.1 | 11 | 1000 | |||
XM_017027984.1 | XP_016883473.1 | 11 | 1106 | |||
XM_017027985.1 | XP_016883474.1 | 4 | 262 | |||
GenBank mRNA | AB037863.1 | - | 10 | 1102 | ||
BC037555.1 | - | 2 | 208 |