Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BAHCC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291324.1 | NP_001278253.1 | 15, 16 | 4684 | ||
XM_011525063.2 | XP_011523365.1 | 16, 17 | 5076 | |||
XM_017024898.1 | XP_016880387.1 | 15, 16 | 4735 | |||
XM_017024899.1 | XP_016880388.1 | 14, 15 | 4641 | |||
GenBank mRNA | AB040880.1 | - | 11, 12 | 2435 |