Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM234B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020853.1 | NP_065904.1 | 12 | 1750 | ||
XM_017019706.1 | XP_016875195.1 | 14 | 2061 | |||
GenBank mRNA | AB040900.1 | - | 9 | 1157 | ||
AK125278.1 | - | 12 | 1750 | |||
AK294732.1 | - | 6 | 762 | |||
BC039267.1 | - | 12 | 1739 | 5274878 | ||
BC132772.1 | - | 12 | 1847 | 40146794 | ||
BC132774.1 | - | 12 | 1847 | 40146796 | ||
CR936647.1 | - | 12 | 1741 |