Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM234B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020853.1 | NP_065904.1 | 4 | 612 | ||
XM_017019706.1 | XP_016875195.1 | 6 | 923 | |||
GenBank mRNA | AB040900.1 | - | 1 | 19 | ||
AK125278.1 | - | 4 | 612 | |||
BC039267.1 | - | 4 | 601 | 5274878 | ||
BC132772.1 | - | 4 | 709 | 40146794 | ||
BC132774.1 | - | 4 | 709 | 40146796 | ||
CR936647.1 | - | 4 | 603 |