Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGAP23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199417.1 | NP_001186346.1 | 2 | 93 | ||
XM_006721991.1 | XP_006722054.1 | 2 | 134 | |||
XM_006721992.3 | XP_006722055.1 | 2 | 121 | |||
XM_006721993.3 | XP_006722056.1 | 2 | 120 | |||
XM_006721994.3 | XP_006722057.1 | 2 | 119 | |||
XM_011525073.1 | XP_011523375.1 | 2 | 53 | |||
XM_011525074.2 | XP_011523376.1 | 2 | 152 | |||
XM_011525076.2 | XP_011523378.1 | 2 | 119 | |||
XM_011525077.2 | XP_011523379.1 | 2 | 119 | |||
XM_017024902.1 | XP_016880391.1 | 2 | 119 | |||
GenBank mRNA | CR746382.1 | - | 2 | 220 |