Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAMSAP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080429.2 | NP_001073898.1 | 13 | 1856 | ||
NM_020902.1 | NP_065953.1 | 11 | 1775 | |||
XM_005272491.3 | XP_005272548.1 | 12 | 1792 | |||
XM_011528154.2 | XP_011526456.1 | 13 | 1775 | |||
XM_017027036.1 | XP_016882525.1 | 13 | 1829 | |||
GenBank mRNA | AB040976.1 | - | 4 | 678 | ||
AL833927.1 | - | 1 | 458 | |||
BC020431.1 | - | 13 | 1699 |