Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBRSL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142641.1 | NP_001136113.1 | 14 | 1978 | ||
XM_005266171.3 | XP_005266228.1 | 17 | 2209 | |||
XM_005266173.3 | XP_005266230.1 | 17 | 2204 | |||
XM_005266175.3 | XP_005266232.1 | 16 | 2143 | |||
XM_005266176.3 | XP_005266233.1 | 16 | 2085 | |||
XM_005266177.3 | XP_005266234.1 | 17 | 2078 | |||
XM_011534803.2 | XP_011533105.1 | 17 | 2268 | |||
XM_011534804.2 | XP_011533106.1 | 17 | 2265 | |||
XM_011534805.2 | XP_011533107.1 | 16 | 2199 | |||
XM_011534806.2 | XP_011533108.1 | 16 | 2202 | |||
XM_011534807.2 | XP_011533109.1 | 16 | 2144 | |||
XM_011534808.2 | XP_011533110.1 | 17 | 2137 | |||
XM_011534809.2 | XP_011533111.1 | 16 | 2080 | |||
XM_011534810.2 | XP_011533112.1 | 17 | 2142 | |||
XM_011534812.2 | XP_011533114.1 | 18 | 2090 | |||
GenBank mRNA | AB046765.2 | - | 14 | 1749 | ||
AF490258.1 | - | 7 | 844 |