Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FANCM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308134.1 | NP_001295063.1 | 2, 3 | 780 | ||
NM_020937.3 | NP_065988.1 | 2, 3 | 780 | |||
XM_011537034.2 | XP_011535336.1 | 2, 3 | 779 | |||
XM_017021523.1 | XP_016877012.1 | 2, 3 | 779 | |||
XM_017021527.1 | XP_016877016.1 | 2, 3 | 726 | |||
GenBank mRNA | AK093422.1 | - | 2, 3 | 779 | ||
BC036056.1 | - | 2, 3 | 781 | |||
DQ140356.1 | - | 2, 3 | 787 |