Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DENND1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020946.1 | NP_065997.1 | 2, 3 | 320 | ||
NM_024820.2 | NP_079096.2 | 2, 3 | 320 | |||
XM_005252109.2 | XP_005252166.1 | 2, 3 | 169 | |||
XM_005252111.4 | XP_005252168.1 | 2, 3 | 1136 | |||
XM_005252113.4 | XP_005252170.1 | 2, 3 | 1125 | |||
XM_006717195.3 | XP_006717258.1 | 2, 3 | 1137 | |||
XM_011518882.2 | XP_011517184.1 | 2, 3 | 1139 | |||
XM_011518883.2 | XP_011517185.1 | 2, 3 | 1140 | |||
XM_011518884.2 | XP_011517186.1 | 2, 3 | 1140 | |||
XM_011518886.2 | XP_011517188.1 | 2, 3 | 1137 | |||
XM_011518887.2 | XP_011517189.1 | 2, 3 | 1128 | |||
XM_017014948.1 | XP_016870437.1 | 2, 3 | 169 | |||
XM_017014949.1 | XP_016870438.1 | 2, 3 | 1140 | |||
XM_017014950.1 | XP_016870439.1 | 2, 3 | 1139 | |||
XM_017014951.1 | XP_016870440.1 | 2, 3 | 163 | |||
GenBank mRNA | AK024782.1 | - | 2, 3 | 308 | ||
AK225104.1 | - | 2, 3 | 308 | |||
BC028061.1 | - | 2, 3 | 160 | |||
BC039703.1 | - | 2, 3 | 173 | 4999484 |