Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WFDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282466.1 | NP_001269395.1 | 5 | 841 | ||
NM_001282467.1 | NP_001269396.1 | 5 | 838 | |||
NM_021197.3 | NP_067020.2 | 5 | 841 | |||
XM_005256079.3 | XP_005256136.1 | 4 | 752 | |||
GenBank mRNA | AF169631.1 | - | 5 | 729 | ||
AF302109.1 | - | 5 | 754 | |||
AK075061.1 | - | 5 | 701 | |||
AL713785.1 | - | 2 | 3503 | |||
BC029159.1 | - | 5 | 647 | 5186156 | ||
BQ670532.1 | - | 5 | 461 | |||
BQ671964.1 | - | 5 | 461 | |||
CD519223.1 | - | 5 | 755 |