Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM60A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135811.1 | NP_001129283.1 | 7 | 2178 | ||
NM_001135812.1 | NP_001129284.1 | 6 | 2074 | |||
NM_021238.2 | NP_067061.1 | 7 | 2174 | |||
XM_011520803.2 | XP_011519105.1 | 6 | 2176 | |||
XM_017019758.1 | XP_016875247.1 | 7 | 2242 | |||
XM_017019759.1 | XP_016875248.1 | 8 | 2288 | |||
XM_017019760.1 | XP_016875249.1 | 8 | 2293 | |||
XM_017019761.1 | XP_016875250.1 | 7 | 2223 | |||
XM_017019762.1 | XP_016875251.1 | 7 | 2140 | |||
GenBank mRNA | AF212220.1 | - | 6 | 2032 | ||
AK026932.1 | - | 6 | 1984 | |||
AL157432.1 | - | 7 | 2132 | |||
BC004462.1 | - | 3 | 1591 | |||
BX648630.1 | - | 5 | 2169 |