Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CACNG6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031897.2 | NP_114103.2 | 2 | 938 | ||
NM_145814.1 | NP_665813.1 | 4 | 1151 | |||
NM_145815.1 | NP_665814.1 | 3 | 1013 | |||
NR_102308.1 | 3 | 141 | ||||
XM_006723316.3 | XP_006723379.1 | 5 | 1817 | |||
GenBank mRNA | AF288386.1 | - | 4 | 691 | ||
AF361352.1 | - | 4 | 561 | |||
BC021552.1 | - | 4 | 1151 | 3357889 | ||
BX281606.1 | - | 3 | 141 |