Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLEKHA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021623.1 | NP_067636.1 | 8 | 911 | ||
XM_011544605.2 | XP_011542907.1 | 8 | 796 | |||
XM_011544606.2 | XP_011542908.1 | 8 | 847 | |||
XM_011544607.2 | XP_011542909.1 | 8 | 809 | |||
XM_017013718.1 | XP_016869207.1 | 7 | 662 | |||
XM_017013719.1 | XP_016869208.1 | 7 | 632 | |||
XM_017013720.1 | XP_016869209.1 | 7 | 747 | |||
GenBank mRNA | AA985353.1 | - | 4 | 224 | ||
AF286164.1 | - | 8 | 836 | |||
AK098787.1 | - | 8 | 912 | |||
AK291842.1 | - | 8 | 855 | |||
AK296068.1 | - | 7 | 704 | |||
AK309020.1 | - | 1 | 28 | |||
DC332022.1 | - | 1 | 28 |