Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDH26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021810.4 | NP_068582.2 | 5 | 462 | ||
NM_177980.2 | NP_817089.1 | 17 | 2565 | |||
XM_011528969.2 | XP_011527271.1 | 17 | 2571 | |||
XM_011528970.2 | XP_011527272.1 | 11 | 1587 | |||
XM_017027994.1 | XP_016883483.1 | 17 | 3234 | |||
XM_017027995.1 | XP_016883484.1 | 16 | 3166 | |||
XM_017027996.1 | XP_016883485.1 | 17 | 3136 | |||
XM_017027997.1 | XP_016883486.1 | 15 | 3112 | |||
XM_017027998.1 | XP_016883487.1 | 17 | 3232 | |||
XM_017027999.1 | XP_016883488.1 | 16 | 3163 | |||
XM_017028000.1 | XP_016883489.1 | 16 | 2404 | |||
XM_017028001.1 | XP_016883490.1 | 15 | 3109 | |||
XM_017028002.1 | XP_016883491.1 | 17 | 3233 | |||
GenBank mRNA | AF169690.1 | - | 7 | 906 | ||
AK055202.1 | - | 17 | 2554 | |||
AK074477.1 | - | 5 | 450 | |||
BC062570.1 | - | 3 | 339 | |||
BC136306.1 | - | 17 | 2548 | 9020293 | ||
BC143931.1 | - | 17 | 2548 |