Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDH26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021810.4 | NP_068582.2 | 6 | 684 | ||
NM_177980.2 | NP_817089.1 | 18 | 2787 | |||
XM_017027998.1 | XP_016883487.1 | 18 | 3454 | |||
XM_017027999.1 | XP_016883488.1 | 17 | 3385 | |||
XM_017028001.1 | XP_016883490.1 | 16 | 3331 | |||
GenBank mRNA | AA454986.1 | - | 1 | 413 | ||
AK055202.1 | - | 18 | 2776 | |||
AK074477.1 | - | 6 | 672 | |||
BC062570.1 | - | 4 | 561 | |||
BC136306.1 | - | 18 | 2770 | 9020293 | ||
BC143931.1 | - | 18 | 2770 |