Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TINAGL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204415.1 | NP_001191344.1 | 3, 4 | 202 | ||
NM_022164.2 | NP_071447.1 | 4, 5 | 594 | |||
XM_005271106.2 | XP_005271163.1 | 4, 5 | 594 | |||
XM_005271107.2 | XP_005271164.1 | 3, 4 | 239 | |||
XM_011541946.1 | XP_011540248.1 | 2, 3 | 306 | |||
GenBank mRNA | AB050716.1 | - | 4, 5 | 544 | ||
AF205436.1 | - | 4, 5 | 520 | |||
AF236150.1 | - | 4, 5 | 522 | |||
AK075398.1 | - | 4, 5 | 567 | |||
AK292770.1 | - | 4, 5 | 568 | |||
AK301070.1 | - | 4, 5 | 604 | |||
AY358421.1 | - | 4, 5 | 567 | |||
BC009048.1 | - | 2, 3 | 125 | |||
BC064633.1 | - | 4, 5 | 552 | 5750149 | ||
BM923842.1 | - | 3, 4 | 202 |