Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TINAGL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204414.1 | NP_001191343.1 | 4 | 597 | ||
NM_001204415.1 | NP_001191344.1 | 4 | 298 | |||
NM_022164.2 | NP_071447.1 | 5 | 690 | |||
XM_005271106.2 | XP_005271163.1 | 5 | 690 | |||
XM_005271107.2 | XP_005271164.1 | 4 | 335 | |||
XM_011541946.1 | XP_011540248.1 | 3 | 402 | |||
GenBank mRNA | AB050716.1 | - | 5 | 640 | ||
AF205436.1 | - | 5 | 616 | |||
AF236150.1 | - | 5 | 618 | |||
AF289569.1 | - | 1 | 46 | |||
AK074124.1 | - | 3 | 899 | |||
AK075398.1 | - | 5 | 663 | |||
AK292770.1 | - | 5 | 664 | |||
AK298038.1 | - | 3 | 792 | |||
AK298382.1 | - | 4 | 567 | |||
AK301070.1 | - | 5 | 700 | |||
AY358421.1 | - | 5 | 663 | |||
BC009048.1 | - | 3 | 221 | |||
BC064633.1 | - | 5 | 648 | 5750149 | ||
BM923842.1 | - | 4 | 298 |