Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HS1BP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022460.3 | NP_071905.3 | 5 | 689 | ||
XM_017004696.1 | XP_016860185.1 | 5 | 692 | |||
XM_017004697.1 | XP_016860186.1 | 5 | 692 | |||
XM_017004698.1 | XP_016860187.1 | 5 | 692 | |||
XM_017004699.1 | XP_016860188.1 | 5 | 744 | |||
XM_017004700.1 | XP_016860189.1 | 5 | 788 | |||
XM_017004701.1 | XP_016860190.1 | 5 | 692 | |||
GenBank mRNA | AK123411.1 | - | 1 | 31 | ||
AK314383.1 | - | 5 | 685 | |||
BC050636.1 | - | 5 | 669 | 5193161 | ||
BG753647.1 | - | 2 | 76 | |||
BX354596.2 | - | 5 | 720 |