Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMP25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022468.4 | NP_071913.1 | 1, 2 | 345 | ||
XM_017023561.1 | XP_016879050.1 | 1, 2 | 1063 | |||
GenBank mRNA | AB042328.1 | - | 1, 2 | 277 | ||
AF145442.2 | - | 1, 2 | 132 | |||
AF185270.1 | - | 1, 2 | 132 | |||
AJ239053.1 | - | 1, 2 | 289 | |||
AJ272137.1 | - | 1, 2 | 345 |