Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMP25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022468.4 | NP_071913.1 | 5 | 986 | ||
XM_011522602.2 | XP_011520904.1 | 5 | 723 | |||
XM_011522604.2 | XP_011520906.1 | 4 | 837 | |||
XM_011522605.2 | XP_011520907.1 | 2 | 177 | |||
GenBank mRNA | AB042328.1 | - | 5 | 918 | ||
AF145442.2 | - | 5 | 773 | |||
AF185270.1 | - | 5 | 773 | |||
AJ239053.1 | - | 5 | 930 | |||
AJ272137.1 | - | 5 | 986 |
Gene Symbol: MMP25-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CN367922.1 | - | 1 | 74 |