Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDH24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022478.3 | NP_071923.2 | 10 | 1816 | ||
NM_144985.3 | NP_659422.2 | 9 | 1702 | |||
XM_011537089.1 | XP_011535391.1 | 10 | 3393 | |||
GenBank mRNA | AK025342.1 | - | 2 | 120 | ||
AK289576.1 | - | 9 | 1702 | |||
AL137477.1 | - | 1 | 275 | |||
AY260900.1 | - | 8 | 1535 | |||
AY260901.1 | - | 9 | 1649 | |||
AY358199.1 | - | 9 | 1658 | |||
BX248303.1 | - | 5 | 726 |